Learn More
RETN, Mouse, Clone: 4B11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RETN.
Brand: Abnova H00056729-M24A.200uL
Description
This gene belongs to the family defined by the mouse resistin-like genes. The characteristic feature of this family is the C-terminal stretch of 10 cys residues with identical spacing. The mouse homolog of this protein is secreted by adipocytes, and may be the hormone potentially linking obesity to type II diabetes. [provided by RefSeq
Sequence: TLCSMEEAINERIQEVAGSLIFRAISSIGLECQSVTSRGDLATCPRGFAVTGCTCGSACGSWDVRAETTCHCQCAGMDWTGARCCRV*Specifications
| RETN | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_020415 | |
| RETN | |
| RETN (NP_065148,20 a.a. ∽ 106 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 56729 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4B11 | |
| Mouse monoclonal antibody raised against a partial recombinant RETN. | |
| RETN | |
| ADSF/FIZZ3/MGC126603/MGC126609/RETN1/RSTN/XCP1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.