Learn More
RFC1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RFC1.
Brand: Abnova H00005981-A01.50uL
Description
The protein encoded by this gene is the large subunit of replication factor C, which is a five subunit DNA polymerase accessory protein. Replication factor C is a DNA-dependent ATPase that is required for eukaryotic DNA replication and repair. The protein acts as an activator of DNA polymerases, binds to the 3' end of primers, and promotes coordinated synthesis of both strands. It also may have a role in telomere stability. [provided by RefSeq]
Sequence: MDIRKFFGVIPSGKKLVSETVKKNEKTKSDEETLKAKKGIKEIKVNSSRKEDDFKQKQPSKKKRIIYDSDSESEETLQVKNAKKPPEKLPVSSKPGKISRQDPVTYISETSpecifications
| RFC1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RFC1. | |
| RFC1 | |
| A1/MGC51786/MHCBFB/PO-GA/RECC1/RFC/RFC140 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC051786 | |
| RFC1 | |
| RFC1 (AAH51786, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 5981 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only