missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RICTOR, Mouse, Clone: 7B3, Abnova™
Mouse monoclonal antibody raised against recombinant RICTOR.
Brand: Abnova MAB10313.100uL
This item is not returnable.
View return policy
Description
RICTOR and MTOR (FRAP1; MIM 601231) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth (Sarbassov et al., 2004 [PubMed 15268862]).[supplied by OMIM
Specifications
| RICTOR | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| RICTOR | |
| RICTOR | |
| Recombinant protein corresponding to human RICTOR. | |
| RUO | |
| 253260 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 7B3 | |
| Mouse monoclonal antibody raised against recombinant RICTOR. | |
| In ascites (0.03% sodium azide) | |
| DKFZp686B11164/KIAA1999/MGC39830/mAVO3 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction