Learn More
RIN3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RIN3.
Brand: Abnova H00079890-A01.50uL
Description
RIN3 is a member of the RIN family of Ras interaction-interference proteins, which are binding partners to the RAB5 small GTPases (see RAB5A; MIM 179512) (Kajiho et al., 2003 [PubMed 12972505]).[supplied by OMIM
Sequence: GPPDHPNQPPMMTCERLPCPTAGLGPLREEAMKPGAASSPLQQVPAPPLPAKKNLPTAPPRRRVSERVSLEDQSPGMAAEGDQLSLPPQGTSDGPEDTPSpecifications
| RIN3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RIN3. | |
| RIN3 | |
| DKFZp762H1613/FLJ11700/FLJ22439 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_024832 | |
| RIN3 | |
| RIN3 (NP_079108, 328 a.a. ∽ 426 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 79890 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only