missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rituximab, Mouse, Clone: 2B11-3, Abnova™
Mouse monoclonal antibody raised against F(ab)2 fragment of Rituximab.
Brand: Abnova MAB11132.100ug
This item is not returnable.
View return policy
Specifications
| Rituximab | |
| Monoclonal | |
| Unconjugated | |
| ELISA (2 ug/mL for coating) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein G purification | |
| RUO | |
| This antibody is monovalent. It blocks Rituximab-CD20 interaction in ELISA. | |
| Liquid |
| Blocking Assay, ELISA | |
| 2B11-3 | |
| Mouse monoclonal antibody raised against F(ab)2 fragment of Rituximab. | |
| In TBS, pH 8.0 (0.09% sodium azide) | |
| F(ab)2 fragment of Rituximab. | |
| 100 μg | |
| Primary | |
| Store at 4°C for at least 12 months. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction