missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1/S100B, Mouse, Clone: 4B3, Abnova™
Mouse monoclonal antibody raised against native S100A1/S100B.
Brand: Abnova MAB3031.200ug
This item is not returnable.
View return policy
Specifications
| S100A1/S100B | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| S100/S100-alpha/S100A | |
| Mouse | |
| 200 μg | |
| Primary | |
| This antibody is specific to S100 beta-beta and S100 alpha-beta. | |
| Store at 4°C. | |
| IgG2a |
| Western Blot | |
| 4B3 | |
| Mouse monoclonal antibody raised against native S100A1/S100B. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| S100A1 | |
| Native purified human brain S100A1/S100B. | |
| RUO | |
| 6271/6285 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction