missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A14, Mouse, Clone: 5, Abnova™
Mouse monoclonal antibody raised against full-length recombinant S100A14.
Brand: Abnova MAB9941.100ug
This item is not returnable.
View return policy
Description
S100A14 is a member of a subfamily of proteins related by Ca(2+)-binding motifs to the EF-hand Ca(2+)-binding protein superfamily.[supplied by OMIM
Specifications
| S100A14 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| S100A14 | |
| BCMP84/S100A15 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 57402 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry | |
| 5 | |
| Mouse monoclonal antibody raised against full length recombinant S100A14. | |
| In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
| NM_020672.1 | |
| S100A14 | |
| Recombinant protein corresponding to full length human S100A14. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction