missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMSN1, Mouse, Clone: 1B8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SAMSN1.
Brand: Abnova H00064092-M01A.200uL
This item is not returnable.
View return policy
Description
SAMSN1 is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM (sterile alpha motif) domains (Claudio et al., 2001 [PubMed 11536050]).[supplied by OMIM
Sequence: MLKRKPSNVSEKEKHQKPKRSSSFGNFDRFRNNSLSKPDDSTEAHEGDPTNGSGEQSKTSNNGGGLGKKMRAISWTMKKKVGKKYIKALSEEKDEEDGENAHPYRNSDPVIGTHTEKVSLKASDSMDSLYSGQSSSSGITSCSDGTSNRDSFRLDDDGPYSGPFCGRARVHTDFTPSPYDTDSLKIKKGDIIDIICKTPMGMWTGMLNNKVGNFKFIYVDVISEEEAAPKKIKANRRSNSKKSKTLQEFLERIHLQEYTSTLLLNGYETLEDLKDIKESHLIELNIENPDDRRRLLSAAENFLEEEIIQEQENEPEPLSLSSDISLNKSQLDDCPRDSGCYISSGNSDNGKEDLESENLSDMVHKIIITEPSDSpecifications
| SAMSN1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC029112 | |
| SAMSN1 | |
| SAMSN1 (AAH29112,1 a.a. ∽ 373 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 64092 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1B8 | |
| Mouse monoclonal antibody raised against a full-length recombinant SAMSN1. | |
| SAMSN1 | |
| HACS1/NASH1/SASH2/SH3D6B | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction