Learn More
SAV1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SAV1.
Brand: Abnova H00060485-A01.50uL
Description
WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 2 WW domains and a coiled-coil region. It is ubiquitously expressed in adult tissues. The encoded protein is 94% identical to the mouse protein at the amino acid level. [provided by RefSeq
Sequence: HTAEIPDWLQVYARAPVKYDHILKWELFQLADLDTYQGMLKLLFMKELEQIVKMYEAYRQALLTELENRKQRQQWYAQQHGKNFSpecifications
| SAV1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SAV1. | |
| SAV1 | |
| SAV/WW45/WWP4 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_021818 | |
| SAV1 | |
| SAV1 (NP_068590, 300 a.a. ∽ 383 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 60485 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only