missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Secretory component (free and bound), Mouse, HRP, Clone: NI 194-4 (A89-039), Abnova™
Mouse monoclonal antibody raised against secretory component (free and bound).
Brand: Abnova MAB10076.200ug
This item is not returnable.
View return policy
Specifications
| Secretory component (free and bound) | |
| Monoclonal | |
| HRP | |
| ELISA (1:50) Western Blot (1:100) Immunohistochemistry (1:20) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Lyophilized |
| ELISA, Immunohistochemistry, Western Blot | |
| NI 194-4 (A89-039) | |
| Mouse monoclonal antibody raised against secretory component (free and bound). | |
| Lyophilized from PBS, pH 7.2 | |
| Purified secretory component (free and bound). | |
| RUO | |
| The antiserum does not react with any other component of the human milk or plasma protein as tested. This antiserum has not been tested for cross-reactivity with other species. | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.5mL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction