Learn More
SELL, Mouse, PE, Clone: LT-TD180, Abnova™
Mouse monoclonal antibody raised against native SELL.
Brand: Abnova MAB4452.100Reactions
Description
SELL is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains: one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins. [provided by RefSeq
Specifications
| SELL | |
| Monoclonal | |
| PE | |
| Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user. | |
| SELL | |
| SELL | |
| Native purified SELL from peripheral blood leukocytes. | |
| RUO | |
| 6402 | |
| Human | |
| Liquid |
| Flow Cytometry | |
| LT-TD180 | |
| Mouse monoclonal antibody raised against native SELL. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| CD62L/LAM-1/LAM1/LECAM1/LNHR/LSEL/LYAM1/Leu-8/Lyam-1/PLNHR/TQ1/hLHRc | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| This antibody reacts with CD62L (L-selectin), a 74-95 KDa single chain type I glycoprotein expressed on most peripheral blood B lymphocytes, T lymphocytes, monocytes and granulocytes; it is also present on a subset of NK cells and certain hematopoietic malignant cells. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.