missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINF1, Mouse, Clone: 14K, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of SERPINF1.
Brand: Abnova MAB1712.50ug
This item is not returnable.
View return policy
Description
The PEDF gene is a member of the serpin gene family. Serpins are a group of serine protease inhibitors, some of which have also been reported to exhibit neurotrophic activity.[supplied by OMIM
Specifications
| SERPINF1 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SERPINF1 | |
| SERPINF1 | |
| A synthetic peptide (conjugated with KLH) corresponding to human SERPINF1. | |
| RUO | |
| 5176 | |
| Liquid |
| Western Blot | |
| 14K | |
| Mouse monoclonal antibody raised against synthetic peptide of SERPINF1. | |
| In PBS (0.1% BSA, 0.1% proclin) | |
| EPC-1/PEDF/PIG35 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction