missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sialyl Lewis a, Mouse, Clone: 121SLE, Abnova™
Mouse monoclonal antibody raised against native Sialyl Lewis a.
Brand: Abnova MAB3589.100ug
This item is not returnable.
View return policy
Specifications
| Sialyl Lewis a | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against native Sialyl Lewis a. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Native purified human Sialyl Lewis a. | |
| RUO | |
| This antibody recognizes Sialyl Lewis a containing glycolipids. | |
| Store at 4°C. Do not freeze. | |
| IgM |
| ELISA, Immunohistochemistry | |
| 121SLE | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction