missing translation for 'onlineSavingsMsg'
Learn More

SLC2A10, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16140377
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16140377 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16140377 Supplier Abnova Supplier No. H00081031A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant SLC2A10.

SLC2A10 is a member of the facilitative glucose transporter family, which plays a significant role in maintaining glucose homeostasis.[supplied by OMIM

Sequence: SGPSCLAVPNATGQTGLPGDSGLLQDSSLPPIPRTNEDQREPILSTAKKTKPHPRSGD

Specifications

Antigen SLC2A10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SLC2A10.
Formulation 50% glycerol
Gene SLC2A10
Gene Accession No. NM_030777
Gene Alias ATS/GLUT10/MGC126706
Gene Symbols SLC2A10
Host Species Mouse
Immunogen SLC2A10 (NP_110404, 325 a.a. ∽ 382 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81031
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.