Learn More
SLC2A2, Mouse, Clone: 5D1, Abnova™
Mouse monoclonal antibody raised against SLC2A2.
Brand: Abnova MAB8054.50ug
Description
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. [provided by RefSeq
Specifications
| SLC2A2 | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry (10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| SLC2A2 | |
| SLC2A2 | |
| Glut2-transfected NS0 cells. | |
| RUO | |
| 6514 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
| 5D1 | |
| Mouse monoclonal antibody raised against SLC2A2. | |
| In PBS (40% glycerol, 5% trehalose) | |
| GLUT2 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.