Learn More
SMARCD3, Mouse, Clone: 5G4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SMARCD3.
Brand: Abnova H00006604-M03A.200uL
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined. [provided by RefSeq
Sequence: ISALDSKIHETIESINQLKIQRDFMLSFSRDPKGYVQDLLRSQSRDLKVMTDVAGNPEEERRAEFYHQPWSQEAVSRYFYCKIQQRRQELEQSLVVRNTSpecifications
| SMARCD3 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_001003801 | |
| SMARCD3 | |
| SMARCD3 (NP_001003801,385 a.a. ∽ 483 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6604 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 5G4 | |
| Mouse monoclonal antibody raised against a partial recombinant SMARCD3. | |
| SMARCD3 | |
| BAF60C/CRACD3/MGC111010/Rsc6p | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.