Learn More
STK11, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant STK11.
Brand: Abnova H00006794-A02.50uL
Description
This gene, which encodes a member of the serine/threonine kinase family, regulates cell polarity and functions as a tumor suppressor. Mutations in this gene have been associated with Peutz-Jeghers syndrome, an autosomal dominant disorder characterized by the growth of polyps in the gastrointestinal tract, pigmented macules on the skin and mouth, and other neoplasms. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq
Sequence: PIPPSPDTKDRWRSMTVVPYLEDLHGADEDEDLFDIEDDIIYTQDFTVPGQVPEEEASHNGQRRGLPKAVCMNGTEAAQLSTKSRAEGRAPNPARKACSASSKIRRLSACKQQSpecifications
| STK11 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant STK11. | |
| STK11 | |
| LKB1/PJS | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC007981 | |
| STK11 | |
| STK11 (AAH07981, 321 a.a. ∽ 433 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 6794 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only