Learn More
STS, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant STS.
Brand: Abnova H00000412-A01.50uL
Description
The protein encoded by this gene catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The encoded protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in this gene are known to cause X-linked ichthyosis (XLI). [provided by RefSeq
Sequence: YEIIQQPMSYDNLTQRLTVEAAQFIQRNTETPFLLVLSYLHVHTALFSSKDFAGKSQHGVYGDAVEEMDWSVGQILNLLDELRLANDTLIYFTSDQGAHVEEVSpecifications
| STS | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant STS. | |
| STS | |
| ARSC/ARSC1/ASC/ES/SSDD | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_000351 | |
| STS | |
| STS (NP_000342, 248 a.a. ∽ 350 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 412 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only