missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUPT5H, Mouse, Clone: 1G3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SUPT5H.
Brand: Abnova H00006829-M04A.200uL
This item is not returnable.
View return policy
Description
Sequence: TTDIQVKVRDTYLDTQVVGQTGVIRSVTGGMCSVYLKDSEKVVSISSEHLEPITPTKNNKVKVILGEDREATGVLLSIDGEDGIVRMDLDEQLKILNLRFLGKLLEASpecifications
| SUPT5H | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC024203 | |
| SUPT5H | |
| SUPT5H (AAH24203,981 a.a. ∽ 1087 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6829 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1G3 | |
| Mouse monoclonal antibody raised against a partial recombinant SUPT5H. | |
| SUPT5H | |
| FLJ34157/SPT5/SPT5H | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction