Learn More
TCEB1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant TCEB1.
Brand: Abnova H00006921-A01.50uL
Description
This gene encodes the protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. [provided by RefSeq
Sequence: MDGEEKTYGGCEGPDAMYVKLISSDGHEFIVKREHALTSGTIKAMLSGPGQFAENETNEVNFREIPSHVLSKVCMYFTYKVRYTNSSTEIPEFPIAPEIALELLMAANFLDCSpecifications
| TCEB1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant TCEB1. | |
| TCEB1 | |
| SIII | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC013809 | |
| TCEB1 | |
| TCEB1 (AAH13809, 1 a.a. ∽ 112 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 6921 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only