missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid-stimulating hormone (TSH), Mouse, Clone: 2/2B4 1A8, Abnova™
Mouse monoclonal antibody raised against recombinant Thyroid-stimulating hormone (TSH).
Brand: Abnova MAB3698.1mg
This item is not returnable.
View return policy
Specifications
| Thyroid-stimulating hormone (TSH) | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Radioimmune Assays (RIA) | |
| 2/2B4 1A8 | |
| Mouse monoclonal antibody raised against recombinant Thyroid-stimulating hormone (TSH). | |
| In BBS | |
| Recombinant protein corresponding to human thyroid-stimulating hormone. | |
| RUO | |
| This antibody recognizes natural and recombinant human Thyroid Stimulating Hormone. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction