Learn More
TIMM13, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant TIMM13.
Brand: Abnova H00026517-A01.50uL
Description
This gene encodes a translocase with similarity to yeast mitochondrial proteins that are involved in the import of metabolite transporters from the cytoplasm and into the mitochondrial inner membrane. The encoded protein and the TIMM8a protein form a 70kDa complex in the intermembrane space. This gene is in a head-to-tail orientation with the gene for lamin B2. [provided by RefSeq
Sequence: MEGGFGSDFGGSGSGKLDPGLIMEQVKVQIAVANAQELLQRMTDKCFRKCIGKPGGSLDNSEQKCIAMCMDRYMDAWNTVSRAYNSRLQRERANMSpecifications
| TIMM13 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant TIMM13. | |
| TIMM13 | |
| TIM13/TIM13B/TIMM13A/TIMM13B/ppv1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC008607 | |
| TIMM13 | |
| TIMM13 (AAH08607, 1 a.a. ∽ 95 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 26517 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only