Learn More
TIMP4, Mouse, HRP, Clone: SB30c, Abnova™
Mouse monoclonal antibody raised against recombinant TIMP4.
Brand: Abnova MAB5896.1mL
Description
This gene belongs to the TIMP gene family. The proteins encoded by this gene family are inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix. The secreted, netrin domain-containing protein encoded by this gene is involved in regulation of platelet aggregation and recruitment and may play role in hormonal regulation and endometrial tissue remodeling. [provided by RefSeq
Specifications
| TIMP4 | |
| Monoclonal | |
| Lot specific | |
| Mouse monoclonal antibody raised against recombinant TIMP4. | |
| In PBS, pH 7.4 (50% glycerol) | |
| TIMP4 | |
| Recombinant protein corresponding to human TIMP4. | |
| RUO | |
| 7079 | |
| Human | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
| SB30c | |
| HRP | |
| The optimal working dilution should be determined by the end user. | |
| TIMP4 | |
| Mouse | |
| 1 mL | |
| Primary | |
| human TIMP-4. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.