Learn More
TLR5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TLR5.
Brand: Abnova H00007100-A01.50uL
Description
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene product is expressed in myelomonocytic cells, and recognizes bacterial flagellin, a principal component of bacterial flagella and a virulence factor. The activation of this receptor mobilizes the nuclear factor NF-kappaB and stimulates tumour necrosis factor-alpha production. [provided by RefSeq
Sequence: LPPGVFSHLTALRGLSLNSNRLTVLSHNDLPANLEILDISRNQLLAPNPDVFVSLSVLDITHNKFICECELSTFINWLNHTNVTIAGPPADIYCVYPDSFSGVSLFSLSTEGCDEEEVSpecifications
| TLR5 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TLR5. | |
| TLR5 | |
| FLJ10052/MGC126430/MGC126431/SLEB1/TIL3 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003268 | |
| TLR5 | |
| TLR5 (NP_003259, 517 a.a. ∽ 634 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 7100 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only