Learn More
TLR3, Mouse, Clone: 44C143, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of TLR8.
Brand: Abnova MAB0089.100ug
Description
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is predominantly expressed in lung and peripheral blood leukocytes, and lies in close proximity to another family member, TLR7, on chromosome X. [provided by RefSeq
Specifications
| TLR3 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| TLR8 | |
| TLR8 | |
| A synthetic peptide corresponding to amino acids 750-850 of human TLR8. | |
| RUO | |
| 51311 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| 44C143 | |
| Mouse monoclonal antibody raised against synthetic peptide of TLR8. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| CD288/MGC119599/MGC119600 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.