Learn More
TNNI3, Mouse, Clone: 3A10A12, 5C3A7, Abnova™
Mouse monoclonal antibody raised against partial recombinant TNNI3.
Brand: Abnova MAB8299.100uL
Description
Troponin I (TnI), along with troponin T (TnT) and troponin C (TnC), is one of 3 subunits that form the troponin complex of the thin filaments of striated muscle. TnI is the inhibitory subunit; blocking actin-myosin interactions and thereby mediating striated muscle relaxation. The TnI subfamily contains three genes: TnI-skeletal-fast-twitch, TnI-skeletal-slow-twitch, and TnI-cardiac. This gene encodes the TnI-cardiac protein and is exclusively expressed in cardiac muscle tissues. Mutations in this gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM). [provided by RefSeq
Specifications
| TNNI3 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:200-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:1000) Immunohistochemistry (Frozen sections) (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end | |
| TNNI3 | |
| TNNI3 | |
| Recombinant protein corresponding to truncated human TNNI3. | |
| RUO | |
| 7137 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| 3A10A12, 5C3A7 | |
| Mouse monoclonal antibody raised against partial recombinant TNNI3. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| CMD2A/CMH7/MGC116817/RCM1/TNNC1/cTnI | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.