missing translation for 'onlineSavingsMsg'
Learn More

TSH, Mouse, Clone: 3A11G12, Abnova™

Product Code. 16043916
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16043916 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16043916 Supplier Abnova Supplier No. MAB1600.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against full-length native TSH.

Thyrotropin-stimulating hormone (TSH) is a noncovalently linked glycoprotein heterodimer and is part of a family of pituitary hormones containing a common alpha subunit (TSHA; see MIM 118850) and a unique beta subunit that confers specificity.[supplied by OMIM

Specifications

Antigen TSH
Applications ELISA
Classification Monoclonal
Clone 3A11G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native TSH.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide)
Gene TSHB
Gene Alias CHNG4/TSH-BETA
Gene Symbols TSHB
Host Species Mouse
Immunogen Native purified human TSH.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7252
Test Specificity This antibody react with TSH and shows low cross-reactivity (less than 0.01%) against LH. No cross-reactivity with hCG and FSH.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.