missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB3, Mouse, Clone: TU-20, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of TUBB3.
Brand: Abnova MAB0907.100ug
This item is not returnable.
View return policy
Description
Sequence: ESESQGPKSpecifications
| TUBB3 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| TUBB3 | |
| TUBB3 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 441-448 of human TUBB3. | |
| RUO | |
| 10381 | |
| Human | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| TU-20 | |
| Mouse monoclonal antibody raised against synthetic peptide of TUBB3. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| MC1R/TUBB4/beta-4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody recognizes neuron-specific human betaIII-tubulin. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction