Learn More
TUBG1, Mouse, Clone: TU-32, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of TUBG1.
Brand: Abnova MAB3661.100ug
Description
This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7. [provided by RefSeq
Sequence: EYHAATRPDYISWGTQSpecifications
| TUBG1 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| TUBG1 | |
| TUBG1 | |
| A synthetic peptide corresponding to amino acids 434-449 of human TUBG1. | |
| RUO | |
| 7283 | |
| Human | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| TU-32 | |
| Mouse monoclonal antibody raised against synthetic peptide of TUBG1. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| TUBG/TUBGCP1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody recognizes an epitope (amino acids 434-449 in human) within C-terminus of TUBG1, a 48 KDa structural constituent of cytoskeleton and microtubule organizing center (MTOC). | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.