missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin-alpha, Mouse, FITC, Clone: TU-01, Abnova™
Mouse monoclonal antibody raised against native tubulin-alpha.
Brand: Abnova MAB5027.100ug
This item is not returnable.
View return policy
Specifications
| Tubulin-alpha | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against native tubulin-alpha. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Native purified porcine tubulin-alpha. | |
| RUO | |
| This antibody recognizes the defined epitope (aa 65-97) on N-terminal structural domain of alpha-tubulin. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunofluorescence | |
| TU-01 | |
| FITC | |
| Immunofluorescence (1:50) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Pig | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction