Learn More
UBB, Mouse, Clone: 3C12, Abnova™
Mouse monoclonal antibody raised against recombinant UBB.
Brand: Abnova MAB10342.100ug
Description
This gene encodes ubiquitin, one of the most conserved proteins known. Ubiquitin is required for ATP-dependent, nonlysosomal intracellular protein degradation of abnormal proteins and normal proteins with a rapid turnover. Ubiquitin is covalently bound to proteins to be degraded, and presumably labels these proteins for degradation. Ubiquitin also binds to histone H2A in actively transcribed regions but does not cause histone H2A degradation, suggesting that ubiquitin is also involved in regulation of gene expression. This gene consists of three direct repeats of the ubiquitin coding sequence with no spacer sequence. Consequently, the protein is expressed as a polyubiquitin precursor with a final amino acid after the last repeat. Aberrant form of this protein has been noticed in patients with Alzheimer's and Down syndrome. [provided by RefSeq]
Specifications
| UBB | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| UBB | |
| UBB | |
| Recombinant protein corresponding to human UBB. | |
| RUO | |
| 7314 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 3C12 | |
| Mouse monoclonal antibody raised against recombinant UBB. | |
| In PBS (0.5% protein stabilizer, 0.05% sodium azide) | |
| FLJ25987/MGC8385 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.