missing translation for 'onlineSavingsMsg'
Learn More

UCP1, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16187245
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16187245 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16187245 Supplier Abnova Supplier No. H00007350A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant UCP1.

Mitochondrial uncoupling proteins (UCP) are members of the family of mitochondrial anion carrier proteins (MACP). UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is expressed only in brown adipose tissue, a specialized tissue which functions to produce heat. [provided by RefSeq]

Sequence: PVDVVKTRFINSPPGQYKSVPNCAMKVFTNEGPTAF

Specifications

Antigen UCP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant UCP1.
Formulation 50% glycerol
Gene UCP1
Gene Accession No. NM_021833
Gene Alias SLC25A7/UCP
Gene Symbols UCP1
Host Species Mouse
Immunogen UCP1 (NP_068605, 232 a.a. ∽ 267 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7350
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.