Learn More
VCAM1, Mouse, Biotin, Clone: STA, Abnova™
Mouse monoclonal antibody raised against VCAM1.
Brand: Abnova MAB10166.100ug
Description
This gene is a member of the Ig superfamily and encodes a cell surface sialoglycoprotein expressed by cytokine-activated endothelium. This type I membrane protein mediates leukocyte-endothelial cell adhesion and signal transduction, and may play a role in the development of artherosclerosis and rheumatoid arthritis. Two alternatively spliced transcripts encoding different isoforms have been described for this gene. [provided by RefSeq
Specifications
| VCAM1 | |
| Monoclonal | |
| Biotin | |
| ELISA Flow Cytometry (4-6 ug/mL) Immunohistochemistry Immunoprecipitation The optimal working dilution should be determined by the end user. | |
| VCAM1 | |
| VCAM1 | |
| Human DS6 T cell line. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| STA | |
| Mouse monoclonal antibody raised against VCAM1. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CD106/DKFZp779G2333/INCAM-100/MGC99561 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 7412 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.