missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSTM1, Mouse, Clone: 2E12, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant VSTM1.
Brand: Abnova H00284415-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MTAEFLSLLCLGLCLGYEDEKKNEKPPKPSLHAWPSSVVEAESNVTLKCQAHSQNVTFVLRKVNDSGYKQEQSSAENEAEFPFTDLKPKDAGRYFCAYKTTASHEWSESSEHLQLVVTDKHDELEAPSMKTDTRTIFVAIFSCISILLLFLSVFIIYRCSQHSELRERKGREGESpecifications
| VSTM1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC100942.2 | |
| VSTM1 | |
| VSTM1 (AAI00943.1,1 a.a. ∽ 174 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2E12 | |
| Mouse monoclonal antibody raised against a full-length recombinant VSTM1. | |
| VSTM1 | |
| MGC119160/MGC119161/UNQ3033 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 284415 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction