Learn More
VWF, Mouse, Clone: VWF635, Abnova™
Mouse monoclonal antibody raised against partial recombinant VWF.
Brand: Abnova MAB11356.100ug
Description
The glycoprotein encoded by this gene functions as both an antihemophilic factor carrier and a platelet-vessel wall mediator in the blood coagulation system. It is crucial to the hemostasis process. Mutations in this gene or deficiencies in this protein result in von Willebrand's disease. An unprocessed pseudogene has been found on chromosome 22. [provided by RefSeq]
Specifications
| VWF | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (1:100-1:200) Immunohistochemistry (1:100-1:200 for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein) Western Blot (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| VWF | |
| VWF | |
| Recombinant protein corresponding to amino acids 845-949 of human VWF. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| VWF635 | |
| Mouse monoclonal antibody raised against partial recombinant VWF. | |
| In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide) | |
| F8VWF/VWD | |
| Mouse | |
| Protein A/G affinity chromatography | |
| RUO | |
| 7450 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.