Learn More
WSB2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant WSB2.
Brand: Abnova H00055884-A01.50uL
Description
This gene encodes a member of the WD-protein subfamily. The encoded protein contains five WD-repeats spanning most of the protein and an SOCS box in the C-terminus. The SOCS box may act as a bridge between specific substrate-binding domains and E3 ubiquitin protein ligases. [provided by RefSeq
Sequence: MEAGEEPLLLAELKPGRPHQFDWKSSCETWSVAFSPDGSWFAWSQGHCIVKLIPWPLEEQFIPKGFEAKSRSSKNETKGRGSPKEKTLDCGQIVWGLAFSPWPSPSpecifications
| WSB2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant WSB2. | |
| WSB2 | |
| MGC10210/SBA2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC015887 | |
| WSB2 | |
| WSB2 (AAH15887, 1 a.a. ∽ 105 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 55884 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only