missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3HAV1L, Mouse, Clone: 2D12, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ZC3HAV1L.
Brand: Abnova H00092092-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MAEPTVCSFLTKVLCAHGGRMFLKDLRGHVELSEARLRDVLQRAGPERFLLQEVETQEGLGDAEAEAAAGAVGGGGTSAWRVVAVSSVRLCARYQRGECQACDQLHFCRRHMLGKCPNRDCWSTCTLSHDIHTPVNMQVLKSHGLFGLNENQLRILLLQNDPCLLPEVCLLYNKGEALYGYCNLKDKCNKFHVCKSFVKGECKLQTCKRSHQLIHAASLKLLQDQGLNIPSVVNFQIISTYKHIKLHKMLENTDNSSPSTEHSQGLEKQGVHAAGAAEAGPLASVPAQSAKKPCPVSCEKSpecifications
| ZC3HAV1L | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_080660.2 | |
| ZC3HAV1L | |
| ZC3HAV1L (NP_542391.1,1 a.a. ∽ 300 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2D12 | |
| Mouse monoclonal antibody raised against a full-length recombinant ZC3HAV1L. | |
| ZC3HAV1L | |
| C7orf39/MGC14289 | |
| Mouse | |
| Protein A Purification | |
| RUO | |
| 92092 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction