Learn More
ZHX2, Mouse, Clone: 5E2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ZHX2.
Brand: Abnova H00022882-M01A.200uL
Description
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family. [provided by RefSeq
Sequence: MEQYQHQPMADDHGYDAVARKATKPMAESPKNGGDVVPQYYKDPKKLCEEDLEKLVTRVKVGSEPAKDCLPAKPSEATSDRSEGSSRDGQGSDENEESSpecifications
| ZHX2 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_014943 | |
| ZHX2 | |
| ZHX2 (NP_055758,691 a.a. ∽ 788 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 22882 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 5E2 | |
| Mouse monoclonal antibody raised against a partial recombinant ZHX2. | |
| ZHX2 | |
| AFR1/KIAA0854/RAF | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.