missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Artemin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1085
This item is not returnable.
View return policy
Description
Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot.
Specifications
| Artemin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9Z0L2.1 | |
| ARTN | |
| E. coli-derived recombinant mouse Artemin Ala112-Gly224 Accession # Q9Z0L2.1 | |
| 500 μg | |
| Primary | |
| Detects mouse Artemin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Lefty A, recombinant mouse (rm) ART, rmCripto, rhCripto-1, recombinant drosophila DPP, recombinant rat (rr) GDNF, rhLAP, rmLefty-1, rrMIS, rhPersephin, rmNodal, recombinant amphibian TGF-beta 5, rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, rhTGF-alpha , or rhNeurturin is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 185230 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9048 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction