missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Autoimmune Regulator/AIRE Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC6184U-100UG
This item is not returnable.
View return policy
Description
Autoimmune Regulator/AIRE Monoclonal specifically detects Autoimmune Regulator/AIRE in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Autoimmune Regulator/AIRE | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Z0E3 | |
| AIRE | |
| E. coli-derived recombinant mouse AIRE Ser476-Ser552 (predicted) Accession # Q9Z0E3 | |
| RUO | |
| 326 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 609930 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| AIRE, APECED, APECED protein, APECEDAPSI, APS1, APSI, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, autoimmune regulator, autoimmune regulator (APECED protein)10APS1AIRE1, autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy), PGA1 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse AIRE in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human AIRE is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction