missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B220/CD45R Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB1217T-100UG
This item is not returnable.
View return policy
Description
B220/CD45R Monoclonal antibody specifically detects B220/CD45R in Mouse samples. It is validated for Flow Cytometry
Specifications
| B220/CD45R | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5788 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| RA3-6B2 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Abelson mouse leukemia virus-induced pre-B tumor cells | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction