missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B7-H4 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB2154S-100UG
This item is not returnable.
View return policy
Description
B7-H4 Monoclonal specifically detects B7-H4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| B7-H4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q7TSP5 | |
| VTCN1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-H4 Phe29-Pro258 Accession # Q7TSP5 | |
| RUO | |
| 79679 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 297219 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse B7-H4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) B7-1, rmB7-2, rmB7-H1, rmB7-H2, rmB7-H3, recombinant human (rh) B7-H3b, rhB7-H4, or rmPD-L2 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction