missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse C-Reactive Protein/CRP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1829-SP
This item is not returnable.
View return policy
Description
C-Reactive Protein/CRP Polyclonal specifically detects C-Reactive Protein/CRP in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| C-Reactive Protein/CRP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1401 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q91XB3 | |
| CRP | |
| Mouse myeloma cell line NS0-derived recombinant mouse C-Reactive Protein/CRP His20-Ser225 Accession # Q91XB3 | |
| 25 μg | |
| Primary | |
| Detects mouse C-Reactive Protein/CRP in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction