missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Carboxylesterase 2/CES2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5280
This item is not returnable.
View return policy
Description
Carboxylesterase 2/CES2 Polyclonal specifically detects Carboxylesterase 2/CES2 in Mouse samples. It is validated for Western Blot.
Specifications
| Carboxylesterase 2/CES2 | |
| Polyclonal | |
| Unconjugated | |
| Q91WG0 | |
| Ces2c | |
| Mouse myeloma cell line NS0-derived recombinant mouse Carboxylesterase 2/CES2 Gln27-Lys557 (predicted) Accession # Q91WG0 | |
| 100 μg | |
| Primary | |
| Detects mouse Carboxylesterase 2/CES2 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human CES2 is observed and approximately 5% cross-reactivity with recombinant mouse (rm) CES3 and rmCES5 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| carboxylesterase 2 (intestine, liver), carboxylesterase 2EC 3.1.1.1, CE-2, CE-2PCE-2, CES2, CES2A1, EC 3.1.1, EC 3.1.1.84, hCE-2, iCE, intestinal carboxylesterase; liver carboxylesterase-2, PCE-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8824 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction