missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin A/Lysosomal Carboxypeptidase A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1123
This item is not returnable.
View return policy
Description
Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Cathepsin A/Lysosomal Carboxypeptidase A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P16675 | |
| CTSA | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin A/Lysosomal Carboxypeptidase A Ala24-Tyr474 (predicted) Accession # P16675 | |
| 500 μg | |
| Primary | |
| Detects mouse Cathepsin A/Lysosomal Carboxypeptidase A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Cathepsin A or recombinant mouse (rm) DPPIV/CD26 is observed. In Western blots, it recognizes the single chain (55 kDa) form of rmCathepsin A, which can be processed into heavy (32 kDa) and light (20 kDa) chains. It recognizes the heavy chain, but not the light chain. | |
| Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 191430 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, CTSA, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, Lysosomal Carboxypeptidase A, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5476 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction