missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin X/Z/P Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF1033
This item is not returnable.
View return policy
Description
Cathepsin X/Z/P Polyclonal specifically detects Cathepsin X/Z/P in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, Immunocytochemistry.
Specifications
| Cathepsin X/Z/P | |
| Polyclonal | |
| Unconjugated | |
| Q9WUU7 | |
| Ctsz | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin X/Z/P Ala23-Val306 Accession # Q9WUU7 | |
| 100 μg | |
| Primary | |
| Detects mouse Cathepsin X/Z/P in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Immunoprecipitation, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2.0 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Cathepsin P, Cathepsin Z, CTSX, CTSZ | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1522 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction