Learn More
Mouse CCL2/JE/MCP-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 43 publications
Brand:  R&D Systems AF-479-NA
Product Code. 18022102
  • € 675.00 / 100µg
Description
CCL2/JE/MCP-1 Polyclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL2/JE/MCP-1
Polyclonal
Unconjugated
Q5SVU3
Ccl2
E. coli-derived recombinant mouse CCL2/JE/MCP-1 Gln24-Arg96 Accession # Q5SVU3
100 μg
Primary
Detects mouse CCL2/JE/MCP-1 in ELISAs and Western blots.
Mouse
IgG
Western Blot, Neutralization, ELISA
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.5-2.5 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF
Goat
Affinity Purified
RUO
6347
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.