Learn More
Mouse CCL2/JE/MCP-1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems MAB479R-01M
 View more versions of this product
Product Code. 18148920
  • € 1595.00 / 1mg
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL2/JE/MCP-1
Monoclonal
Unconjugated
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF
Rat
Protein A or G purified from cell culture supernatant
RUO
6347
Mouse
Purified
Flow Cytometry
123616R
Flow Cytometry 0.25 ug/10^6 cells
P10148
Ccl2
E. coli-derived recombinant mouse CCL2 Gln24-Arg96 Accession No. P10148
1 mg
Primary
Detects mouse CCL2 in direct ELISAs.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b