missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL21/6Ckine Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems BAF457
This item is not returnable.
View return policy
Description
CCL21/6Ckine Polyclonal specifically detects CCL21/6Ckine in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| CCL21/6Ckine | |
| Polyclonal | |
| Biotin | |
| P84444 | |
| Ccl21a | |
| E. coli-derived recombinant mouse CCL21/6Ckine (R&D Systems, Catalog # 457-6C) Ser24-Gly133 Accession # P84444 | |
| 50 μg | |
| Primary | |
| Detects mouse CCL21/6Ckine in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with recombinant human 6Ckine, recombinant mouse (rm) Eotaxin-2, rmRANTES, rmMIP-1 alpha , rmMIP-1 beta , rmMIP-3 beta , rmTECK, and rmTARC is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| 6Ckine, 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, exodus-2, member 21, SCYA21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLC, SLCSecondary lymphoid-tissue chemokine, TCA-4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6366 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction