missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL3/MIP-1 alpha APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC450A
This item is not returnable.
View return policy
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CCL3/MIP-1 alpha | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6348 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 39624 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P10855 | |
| CCL3 | |
| E. coli-derived recombinant mouse CCL3/MIP-1 alpha Ala24-Ala92 Accession # P10855 | |
| 100 Tests | |
| Primary | |
| Detects mouse CCL3/MIP-1 alpha in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse CCL4/MIP-1 beta is observed and no cross-reactivity with recombinant human CCL1, 2, 3, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 21, 22, 23, 24, 25, 28, recombinant mouse CCL1, 2, 6, 7, 9/10/MIP-1 gamma , 11, 12, 19, 21, 22, 25, 28, recombinant rat CCL2 or CCL20 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction